Synthware High Vacuum Valve, Teflon® Protected, 90° Metering
Supplier: Kemtech America
S670008
60004-058EA 132.81 USD
60004-058
Synthware High Vacuum Valve, Teflon® Protected, 90° Metering
Valves Vacuum Valves
Viton® O-Rings on PTFE shaft are protected with precision Teflon®wipers. The plug has a machined-in integral PTFE tip O-Ring to make the primary seal.
  • Teflon shaft
  • PTFE covered Viton O-Rings
Valves are designed to provide an excellent vacuum tight seal without exposing any material other than Teflon® to chemicals.