Synthware Round Bottom Flasks with Three Vertical Necks, Kemtech America
Supplier: Kemtech America
F441150 F449950 F441100 F449100 F449900 F444100 F444900 F444400 F441250 F442250 F444250 F441500 F449500 F440500 F444500 F446500 F448500 F444000 F44841L F44341L F44451L F44242L F44842L F44292L F44342L F44542L F44543L F44583L F44245L F44845L F44295L F44345L F44385L F44545L F44585L F44243L F44843L F44293L F44343L F44383L
60002-782EA 75.87 USD
60002-782 60002-786 60002-788 60002-790 60002-792 60002-794 60002-796 60002-798 60002-800 60002-806 60002-808 60002-810 60002-814 60002-816 60002-818 60002-820 60002-822 60002-836 60002-838 60002-842 60002-846 60002-850 60002-852 60002-854 60002-856 60002-860 76264-102 76264-104 76264-106 76264-108 76264-110 76264-112 76264-114 76264-116 76264-118 76264-374 76264-376 76264-378 76264-380 76264-382
Synthware Round Bottom Flasks with Three Vertical Necks, Kemtech America
Flasks Round Bottom Flasks
Round-bottom with [ST] joints.
  • 3 Straight necks
  • Ground glass joints
  • Heavy wall borosilicate construction
  • Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.