Synthware Round-Bottom Flasks with Spherical Joint, Kemtech America
Supplier: Kemtech America
F527500 F527000 F52702L
60002-366EA 42.15 USD
60002-366 60002-368 60002-370
Synthware Round-Bottom Flasks with Spherical Joint, Kemtech America
Flasks Round Bottom Flasks
Heavy wall design.
  • Single Neck
  • Spherical ground glass joint
  • Heavy wall borosilicate construction
  • Various capacities and joint sizes svailable
All sizes are hand-blown from tubing to ensure uniform wall thickness.