Synthware Desiccant Flasks, Kemtech America
Supplier: Kemtech America
D133424
60002-158EA 139.71 USD
60002-158
Synthware Desiccant Flasks, Kemtech America
Desiccators Desiccators, Glass
Choice of either glass or PTFE stopcock
  • Heavy wall borosilicate construction
  • 34/45 ground glass joint
  • Teflon® stopcock
[ST] joint size: 34/45.