Synthware Distillation Apparatus, Large Scale, Kemtech America
Supplier: Kemtech America
D502440
60002-152EA 501.4 USD
60002-152
Synthware Distillation Apparatus, Large Scale, Kemtech America
Distillation Components Distillation Sets
One-piece design with [ST]10/18 thermometer joint and high-efficiency cooling jacket
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Jacketed
  • Large scale applications
Used with 50mm immersion thermometer.
Ordering information: Thermometer is not supplied; contact your Avantor representative for more information.