Synthware Micro Distillation Head, Jacketed Vigreux, Kemtech America
Supplier: Kemtech America
D171014
60002-142EA 428.84 USD
60002-142
Synthware Micro Distillation Head, Jacketed Vigreux, Kemtech America
Distillation Components Distillation Heads
With the Vigreux column vacuum jacketed to improve distillation performance.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Vigreux indentations
  • Designated 10/18 thermometer joint
Used with 60002-132 and a 25 mm immersion thermometer.
Ordering information: Distribution adapter and thermometer are not supplied. For thermometer, contact your Avantor representative.