Synthware Distribution Receiver, Vertical Orientation, Kemtech America
Supplier: Kemtech America
A592440
60002-138EA 82.07 USD
60002-138
Synthware Distribution Receiver, Vertical Orientation, Kemtech America
Distillation Components Distillation Receivers
Rotating distillation receiver allows four fractions to be collected.
  • Vertical orientation
  • Heavy wall borosilicate construction
  • Lower drip tip
  • Hose connection side arm
Vacuum adapter has a vertical orientation.