Synthware Distillation Receiving Sets, Distribution Adapter, Kemtech America
Supplier: Kemtech America
A551420 A551922 A552440 D201414 D201922 D202424
60002-126EA 60.29 USD
60002-126 60002-128 60002-130 60002-132 60002-134 60002-136
Synthware Distillation Receiving Sets, Distribution Adapter, Kemtech America
Distillation Components Distillation Receivers
Rotating cow receiver allows four flasks to be progressively filled.
  • Heavy wall Borosilicate construction
  • Ground glass joint
  • Keck clips included
The distillation receiver set allows you to distill into three separate flasks.
Ordering information: Set components are sold separately.