Synthware Distilling Head for Volatile Solvents, Short Path, Kemtech America
Supplier: Kemtech America
D531024
60002-080EA 419.53 USD
60002-080
Synthware Distilling Head for Volatile Solvents, Short Path, Kemtech America
Distillation Components Distillation Heads
Condenser is a combination of a cold coil and Liebig.
  • Heavy wall Borosilicate construction
  • Ground glass joint
  • Jacketed
  • Microscale
  • Drip tip
  • Designated 10/18 Thermometer joint
Top outer joint is [ST]10/18 for use with a 50 mm immersion thermometer.
Ordering information: Distillation receiver and thermometer are not supplied. For best results, use distillation receiver 60002-064. For thermometer, contact your Avantor representative.