Synthware Short Path Distillation Apparatus with [ST]24/40 Joint, Kemtech America
Supplier: Kemtech America
D261024
60002-078EA 287.77 USD
60002-078
Synthware Short Path Distillation Apparatus with [ST]24/40 Joint, Kemtech America
Distillation Components Distillation Sets
One-piece design for easy set up.
  • Heavy wall borosilicate construction
  • Short path
  • Ground glass joint
  • Jacketed
  • Designated 10/18 thermometer joint
Used with 25 mm immersion thermometer.
Ordering information: Distillation receiver and thermometer are not supplied. For best results, use distillation receiver 60002-064. For thermometer, contact your Avantor representative.