Synthware Distilling Receivers, Kemtech America
Supplier: Kemtech America
D381412
60002-072EA 281.59 USD
60002-072
Synthware Distilling Receivers, Kemtech America
Distillation Components Distillation Receivers
With 135° angle between top outer joint and lower inner joints.
  • Heavy wall Borosilicate construction
  • Ground glass joint
  • Keck clips included
  • graduated receiver tubes included
Supplied with distribution adapter, three 12 ml receiver tubes, and three metal clamps.