Synthware Short Path Distillation Apparatus, Vacuum-Jacketed, Kemtech America
Supplier: Kemtech America
D121014
60002-070EA 495.76 USD
60002-070
Synthware Short Path Distillation Apparatus, Vacuum-Jacketed, Kemtech America
Distillation Components Distillation Sets
Vacuum jacketed design provides more efficiency.
  • Heavy wall borosilicate construction
  • Short path
  • Ground glass joint
  • Jacketed
  • Designated 10/18 thermometer joint
When a thermometer is inserted in the top of this apparatus, the bulb and stem serve as column packing. The close coupled condenser and vacuum connection provide an extremely short condensate travel path. Used with 25mm immersion thermometer and distillation receiver.
Ordering information: Distillation receiver and thermometer are not supplied. For distallation receiver, see 60002-060 series. For thermometer, contact your Avantor representative.