Synthware Chromatography Columns with Beaded Top, Fritted Disc, and PTFE Stopcock
Supplier: Kemtech America
C361720C C361725C C361730C C361740C C362620C C362625C C362630C C362640C C363225C C363230C C363240C C364620C C364625C C364630C C364640C C366040C C366060C C368030C
60001-696EA 125.48 USD
60001-696 60001-698 60001-700 60001-702 60001-704 60001-706 60001-708 60001-710 60001-714 60001-716 60001-718 60001-720 60001-722 60001-724 60001-726 60001-734 60001-736 60001-742
Synthware Chromatography Columns with Beaded Top, Fritted Disc, and PTFE Stopcock
Chromatography Columns
Chromatography columns have a reinforced beaded top, coarse porosity fritted disc to support column packing, and PTFE stopcock.
  • Heavy wall borosilicate construction
  • Beaded top
  • Teflon® stopcock
  • High capacity reservoir
  • Fritted disc
Chromatography columns are used to separate out individual components of a liquid sample.