Synthware Liebig Condensers
Supplier: Kemtech America
C201120 C209180 C209300 C204100 C204200 C204250 C204300 C204400 C204500
60001-138EA 115.83 USD
60001-138 60001-140 60001-142 60001-144 60001-146 60001-148 60001-150 60001-152 60001-154
Synthware Liebig Condensers
Condensers
With top outer [ST] joint and bottom inner [ST] joint.
  • Liebig style condenser
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Jacketed
Jacketed liebig condenser is perfect for most chemical distillation applications.