Synthware Distilling Column Packing, Kemtech America
Supplier: Kemtech America
D240001
60000-792EA 121.06 USD
60000-792
Synthware Distilling Column Packing, Kemtech America
Distillation Column Packing
Particle-type mini glass coils packing can be poured into any laboratory or pilot scale column.
Easily removed for replacement or cleaning. Size: 4 (int.Ø)×12 mm. Packaging: 500g/pack.