Synthware Gas Washing Bottle with Extra-Large Fritted Disc, Kemtech America
Supplier: Kemtech America
B310350C
60000-760EA 304.85 USD
60000-760
Synthware Gas Washing Bottle with Extra-Large Fritted Disc, Kemtech America
Bottles Gas Washing Bottles
Gas washing bottle with an extra-large diameter (60 mm) fritted disc sealed into the bottom of the bottle to enable increased gas throughput and uniform gas distribution.
  • Porosity: coarse
Height (approx.): 250 mm.
Ordering information: For a complete bottle, the [ST]24/40 top outer joint accepts gas inlet adapter 60000-166 (ordered separately).