Synthware Bubblers with Bottom Hose Connection, Kemtech America
Supplier: Kemtech America
B262910
60000-726EA 118.18 USD
60000-726
Synthware Bubblers with Bottom Hose Connection, Kemtech America
Bubblers
With small ball joints to prevent mineral oil from being sucked back under sudden negative pressure in the system.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Anti backflow design
Bottom hose connection diameter: 10 mm. Length: 157 mm.