Synthware Kjeldahl Trap Adapters, Kemtech America
Supplier: Kemtech America
A172440
60000-712EA 154.61 USD
60000-712
Synthware Kjeldahl Trap Adapters, Kemtech America
Distillation Components Distillation Adapters
Kjeldahl trap adapters feature two [ST] 24/40 joints.
  • Used to catch splashes
  • With two joints
The span between the centers of joints is approximately 200 mm.