Synthware Three-Arm Connecting Adapters, One Inlet, Kemtech America
Supplier: Kemtech America
A101420 A102440
60000-674EA 90.07 USD
60000-674 60000-676
Synthware Three-Arm Connecting Adapters, One Inlet, Kemtech America
Adapters Adapters, Conical Ground Joints
The three-arm design of this adapter allows for the assembly of three vessels.
  • Three arms
  • One inlet
Connecting adapter with one female joint on top and two male joints below.