Synthware Connecting Adapters with Three Arms, Two Inlet, Kemtech America
Supplier: Kemtech America
A331420 A332440
60000-666EA 84.47 USD
60000-666 60000-668
Synthware Connecting Adapters with Three Arms, Two Inlet, Kemtech America
Adapters Adapters, Conical Ground Joints
The three-arm design of this adapter allows for the assembly of three vessels.
  • Three arms
  • Two inlets
Connecting adapter with two female joints on top and a male joint below.