Synthware Distillation Adapters, Kemtech America
Supplier: Kemtech America
A362410
60000-656EA 115.92 USD
60000-656
Synthware Distillation Adapters, Kemtech America
Distillation Components Distillation Adapters
Distillation adapter accommodates a 76 mm immersion thermometer.
  • Accommodates thermometer
  • Thermometer not included
Features a [ST] 10/18 joint on the vertical side tube for a 76 mm immersion thermometer.
Ordering information: Thermometer is not supplied; contact your Avantor representative for more information.