Synthware Distillation Connecting Adapters, Vigreux, Kemtech America
Supplier: Kemtech America
A131024
60000-644EA 95.05 USD
60000-644
Synthware Distillation Connecting Adapters, Vigreux, Kemtech America
Distillation Components Distillation Adapters
Distillation connecting adapter accommodates a 76 mm immersion thermometer.
  • Accommodates thermometer
  • Thermometer not included
Top [ST] joint is 10/18. Side [ST] joint is 24/40. Other [ST] joint is 24/40.
Ordering information: Thermometer is not supplied; contact your Avantor representative for more information.