Synthware Inlet Adapters with Hose Connection, Kemtech America
Supplier: Kemtech America
A431420 A431922 A432440
60000-334EA 51.73 USD
60000-334 60000-336 60000-338
Synthware Inlet Adapters with Hose Connection, Kemtech America
Adapters Adapters, Joints with Tube Connectors
This inlet adapter features a hose connection, vacuum take-off and drip tip.
  • Vacuum take-off
  • Drip tip
  • With hose connection
Inlet vacuum adapter features a vacuum port on the drain side a drip tip