Synthware Thermometer Adapter, Kemtech America
Supplier: Kemtech America
A162440
60000-288EA 94.33 USD
60000-288
Synthware Thermometer Adapter, Kemtech America
Adapters Adapters, Joints with Tube Connectors
This adapter features a thermometer joint that is angled for use in a multiple neck flask.
  • Angled joint
  • Accommodates thermometer
The angled thermometer joint is size [ST] 10/30.