Synthware Vacuum Adapters with Outer Joint, Kemtech America
Supplier: Kemtech America
A321420 A322440
60000-212EA 29.03 USD
60000-212 60000-216
Synthware Vacuum Adapters with Outer Joint, Kemtech America
Adapters Adapters, Joints with Tube Connectors
This vacuum adapter with outer joint is ideal for use as a vent or for the introduction of pressure or vacuum.
  • Outer [ST] joint
  • With hose connection
Adapter features a hose size of 8mm O.D.