Synthware Vacuum/Gas Adapter, Ball Joint, Kemtech America
Supplier: Kemtech America
A293520
60000-170EA 46.91 USD
60000-170
Synthware Vacuum/Gas Adapter, Ball Joint, Kemtech America
Adapters Adapters, Spherical Ground Joints
Vacuum/gas adapter with a ball joint.
  • [ST] ball joint
  • With hose connection
Featuring a [ST] 35/20 ball joint and 8mm hose connection.