Synthware Inlet Adapters, Kemtech America
Supplier: Kemtech America
A451420 A451922 A452440
60000-138EA 20.42 USD
60000-138 60000-140 60000-142
Synthware Inlet Adapters, Kemtech America
Adapters Adapters, Conical Ground Joints
Use inlet adapters with gas inlet tubes or plain stem thermometers.
  • Inner [ST] joint
  • One-holed rubber adapter included
Features an inner [ST] joint. Supplied with one-holed rubber adapter.
Ordering information: For a replacement rubber adapter, contact your Avantor representative.