KIMAX® Straight-Bore Stopcock, without Plug, DWK LIFESCIENCES
Supplier: DWK Life Sciences (KIMBLE)
41004G-4
59140-088EA 66.68 USD
59140-088
KIMAX® Straight-Bore Stopcock, without Plug, DWK LIFESCIENCES
Stopcocks
Straight-bore, sandblasted stopcock with a 1:5 taper.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.