Bubble Humidifier Kit, Vyaire Medical
Supplier: Carefusion
002620
59100-108CS 270.71 USD
59100-108
Bubble Humidifier Kit, Vyaire Medical
Ventilators
These humidifier systems feature a dual-purpose reservoir with a snap-open trigger, which makes a round opening for secure attachment of a humidifier adapter.
  • Prefilled with sterile water
  • Latex-free
The oxygen pathway is designed to help prevent particulate from reaching oxygen delivery tubing. An audible relief valve warns if the patient’s tubing becomes occluded. They are designed for convenient shelf storage.