SP Bel-Art Stirring Rod, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37766-0008
59063-001CS 267.74 USD
59063-001
SP Bel-Art Stirring Rod, Polypropylene, Bel-Art Products, a part of SP
Stirrers Stirring Rods
This solid, polypropylene plastic stirring rod with rounded end is useful for mixing chemicals and liquids.
  • Solid polypropylene construction
  • Rounded end design
  • Autoclavable
It is steam autoclavable at 121 °C (250 °F).