Stainless Steel Shaft and Propeller Assembly, Arrow
Supplier: Arrow Engineering
316S-ST-15 316S-ST-18 316S-ST-20 316S-ST-24 316S-ST-30 VPP-2625 VPP-3125 VPP-3625 VPP-4750 VPP-5250 316S-ST-12 VPP-4250
59025-138EA 89.27 USD
59025-138 59025-140 59025-142 59025-144 59025-146 59025-230 59025-232 59025-234 59025-238 59025-240 59025-136 59025-236
Stainless Steel Shaft and Propeller Assembly, Arrow
Stirrers Stirring Tools
Variable pitch Arrow propellers are designed so that user can set the pitch of the paddles at several angles within a range to achieve the best agitation result for your application
Type 316 stainless steel shaft and propeller can withstand corrosive attack by sodium and calcium brines, hypochlorite solutions, phosphoric acid, and sulfurous acid.
Each propeller is factory set at a 30° angle from the horizontal plane unless otherwise stated. Paddles can be set at any angle at the time of ordering at no additional charge.
Ordering information: Contact your Avantor representative for couplings. Three sizes are available to fit your motor shaft: 0.6, 1, and 1.3cm (1/4, 3/8, and 1/2") diameter.