Pulverizer, Waring
Supplier: Waring Products
SS110
58983-093EA 697.94 USD
58983-093
Pulverizer, Waring
Homogenizers Standard Laboratory Blenders
Small stainless steel container with special blade configurations reduces small bones, seeds, grains, nuts, soybeans, tablets, soils and similar materials in seconds
Fits all Waring® laboratory blenders (requires an adapter for 2 and 4 liter blenders). Chrome plated die-cast base. Dry capacity: 75g.
Recommended as a replacement for retail coffee mills, which have a short life span in laboratory applications.
Ordering information: Supplied complete with snap tight lid and stainless steel pulverizing blade assembly.