Seven-Speed Laboratory Blenders, 1 L, Waring
Supplier: Waring Products
7012G
58977-227EA 1453.71 USD
58977-227
Seven-Speed Laboratory Blenders, 1 L, Waring
Homogenizers Standard Laboratory Blenders
Seven speed, eight-button, solid-state SCR (silicon controlled rectifier) control provides speeds from 3500 to 22,000rpm (no-load)
Heat resistant glass or stainless steel container with handle and two-piece vinyl and styrene lid. Motor housing and blade are epoxy coated.
Certifications: UL listed.