Single-Speed Blenders, 1 L, Waring
Supplier: Waring Products
700G 700S
58977-001EA 1213.85 USD
58977-001 58977-023
Single-Speed Blenders, 1 L, Waring
Homogenizers Standard Laboratory Blenders
For rapid pulping or mixing
Removable containers of heat-resistant glass or stainless steel have a 1 L working capacity. The 0.5 hp motor revolves blades at 22000 rpm. Enameled metal base.
Widely used for emulsifying and for blending cosmetics, vitamins, paint and varnish, paper, biologicals, and oils.
Certifications: The 120V models are UL listed.
Ordering information: Supplied complete with two-piece lid.