Whirl-Pak® Round-Bottom Bags for Homogenizer Blenders, Nasco®
Supplier: NASCO SAMPLING LLC
B01239
58976-929CS 849.3 USD
58976-929
Whirl-Pak® Round-Bottom Bags for Homogenizer Blenders, Nasco®
Bags Blender Bags
These bags feature a special round bottom seal designed to minimize sample fragments from getting caught in the corners of the bag during blending.
  • Specially designed for use with homogenizer blenders
It is made from extra-heavy polyethylene and has special features to make sample testing easier and more efficient. Non-sparking tools should be used for handling.