High Torque Overhead Stirrers, Caframo
Supplier: Caframo
BDC6015 BDC1850 BDC1850 BDC3030 BDC3030 BDC6015
77776-348EA 2348.37 USD
CA58960-170 CA58960-180 77776-348 58960-447 77776-354 CA58960-165
High Torque Overhead Stirrers, Caframo
Stirrers Overhead Stirrers
Designed for continuous, high viscosity mixing
These benchtop overhead stirrers are most suitable for highly viscous mixing applications. Other applications include cosmetics and personal care products, inks and coatings, paint, pharmaceuticals, pulp and paper, pilot plants, adhesives, and epoxies.
Brushless, sparkless DC motor and two speed transmission deliver constant performance under changing conditions. Speed is maintained through electronic control as product viscosity changes. Stirrers are built with a totally enclosed metal housing, through shaft, digital display, and chuck guard.
Certifications: CSA, CE marked, RoHS and WEEE.
Ordering information: Clamp, stand, and impellers are available. A 220V model is also available. Units are also supplied with a three-year warranty. Contact Avantor customer service to obtain ordering information. Safety stand includes cast metal base and support rod.