VWR® Disposal Carton for Plastics
Supplier: VWR International
24652-6000
58949-700PK 128.89 USD
58949-700
VWR® Disposal Carton for Plastics
Boxes Waste Disposal Boxes
Disposal boxes are shipped flat and fold into shape easily.
  • Clearly labeled, rigid cardboard carton; includes polypropylene liner to contain spills
  • Dedicated to separating plastics from the waste stream
  • Sized to fit under many benchtops
  • Approximate volume: 63 liters (16.6 gallons)
Entire box can be discarded when filled, ensuring safe handling and ready identification of contents.