VWR® Spinbar® Stir Bars, Octagon, Color-Coded
Supplier: Bel-Art Products, a Part of SP
47751-468 47751-470 37109-6032 37109-6033 47751-476 47751-478 58948-085 58948-087 58949-042 58949-044 58949-046 58949-048 58949-050 58949-051 58949-052 58949-053 58949-054 58949-056 58949-058 58949-060 58949-062 58949-064 58949-066 70670-098
47751-468EA 7.34 USD
47751-468 47751-470 47751-472 47751-474 47751-476 47751-478 58948-085 58948-087 58949-042 58949-044 58949-046 58949-048 58949-050 58949-051 58949-052 58949-053 58949-054 58949-056 58949-058 58949-060 58949-062 58949-064 58949-066 70670-098
VWR® Spinbar® Stir Bars, Octagon, Color-Coded
Stirrers Magnetic Stirring Bars
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert
  • Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
  • Pivot ring aids in reducing friction and chattering
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
Available in red, yellow or blue. All stir bars have a pivot ring with the exception of the 12.7 x 3mm sizes (58949-044, -046, and -050). Autoclavable.
Choose from red, yellow or blue.
Ordering information: Assortment includes six of size 1 x 5/16" (25 x 7mm), 2 each color; Yellow, Red, Blue.