VWR® Spinbar® Micro Stir Bars, Assortment
Supplier: VWR International
58948-069
58948-069CS 1050.65 USD
58948-069
VWR® Spinbar® Micro Stir Bars, Assortment
Stirrers Magnetic Stirring Bars
Think small with a Spinpak® of six metric, flea-sized, PTFE resin-coated magnetic stir bars.
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low-friction coating
  • Durable
  • Inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable
These products are very small, efficient stirring bars.
Ordering information: Includes two of each of 7.9x1.5mm (5/16x1/16"), 7.9x2mm (5/16x1/8"), and 10x3mm (7/16x3/16") sizes.