Staticide® Towelettes and Wipes, ACL Staticide
Supplier: ACL Staticide
SW12
58611-178CS 181.71 USD
58611-178
Staticide® Towelettes and Wipes, ACL Staticide
Wipes Disposable Wipes and Towels Presaturated Wipes
Individually foil-wrapped towelettes and wipes can be used in industrial, commercial, and technical environments for cleaning and static control of nonporous surfaces such as CRT screens, keyboards, and sensitive electronic equipment.
  • Color: White
  • Long-lasting, nontoxic, and biodegradable
Note: Not for use on antiglare/polarized screen filters.
When used often, they prevent dust attraction and build-up. Effective in a relative humidity below 0.15%. Formulated with a blended solution of pure Staticide concentrate, deionized water, and isopropyl alcohol.
Certifications: Meet NFPA-56A and MIL-B-81705B electrostatic decay criteria.