Zero Charge® Mat and Table Top Cleaner, Techspray™
Supplier: ITW Chemtronics
1733-G
58542-414EA 42.07 USD
58542-414
Zero Charge® Mat and Table Top Cleaner, Techspray™
Surface Treatments Floor Treatments
A neutral pH cleaner for general use in production areas, this product removes insulating contaminants such as flux residues, oils, greases, nicotine, dirt, and grime
It eliminates charge while leaving an antistatic agent that helps prevent static charge build-up. Use on conductive mats, table tops, computer and soldering stations, telecommunication equipment, floor mats, electronic instrumentation, and more. Nonabrasive, noncorrosive, nonflammable; will not scratch substrate. Contains no ammonia.