Sponge, Liberty Industries
Supplier: Liberty Industries
50-5128
58540-025EA 15.53 USD
58540-025
Sponge, Liberty Industries
Wipes Sponges
Blown urethane sponges, Class 100 (ISO 5) cleanroom packaged and processed in level 2 bag. Less than 100 particles per cu.ft. 3 oz. Sponges are sold individually.
Certifications: Certified cleanroom cleaned sponge.
Caution: Note: Available in sterile-model numbers with 'S' for sterile. Particle counts available