Shoe Rack, Eagle MHC
Supplier: Eagle Group
SR1460C
58202-446EA 3169.26 USD
58202-446
Shoe Rack, Eagle MHC
Furniture Gowning Racks
Chrome-plated steel rack for shoe storage features wire shelves that allow for 62% laminar air flow.
  • Footplates (included) may be fastened to the floor
Rods and tabs run through the unit to create open compartments for 60 pairs of shoes.
Ordering information: For an electropolished stainless steel rack, contact your Avantor sales representative.