SP Bel-Art Spectro Cuvette Rack, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18515-0000
58020-107CS 288.79 USD
58020-107
SP Bel-Art Spectro Cuvette Rack, Bel-Art Products, a part of SP
Racks Cuvette Racks
This polypropylene rack holds twenty 10 mm light path spectrophotometer cuvettes in two tiers with numbered wells.
  • Polypropylene construction
  • Sloped top improves accessibility to second tier of numbered wells
  • Autoclavable
The wells hold standard cuvettes with a 12.5 mm cross-section. A sloped top makes the second tier more accessible. Steam autoclavable at 121°C (250°F).