Isopropyl Alcohol Solutions, CleanTex™
Supplier: Advantus
CT131
58019-526PK 283.08 USD
58019-526
Isopropyl Alcohol Solutions, CleanTex™
Cleaning Agents Detergents and Surface Cleansers
These solvents are specially formulated to dissolve grease and oils and clean sensitive components, while drying residue free in any cleaning application.
  • Zero ozone depletion
  • Safe for use on delicate substrates
  • For general light-duty cleaning
  • Cleaning of tape heads and transports
  • Precision cleaning of electronic and optical components