PEP™ (Personal Electroporation Pak), BTX™
Supplier: Harvard Apparatus
45-0212
58018-002EA 683.82 USD
58018-002
PEP™ (Personal Electroporation Pak), BTX™
Electroporation Systems Electroporators
The PEP™ (Personal Electroporation Pak) is a self-contained portable cuvette module designed for the ECM® 399
The PEP™'s portability, ease of use, and easy release cuvette ejection pin make it the ideal solution for quick, singular transfections. The PEP™ can be loaded anywhere in the lab then easily transported to the generator. It can be plugged directly into the generator. The PEP™ is compatible with the ECM® 399, ECM® 630, and ECM® 830. Connection cables are not required.