Petri Dish Electrode, BTX®
Supplier: Harvard Apparatus
45-0100
58017-970EA 3365.65 USD
58017-970
Petri Dish Electrode, BTX®
Electrodes Electroporation Electrodes
For use with a 100mm petri dish that functions as the electroporation chamber
Can be used for electroporation of tissues grown in the petri dish. Compatible with BTX ECM® 630 and 830 systems. Electrodes may be rinsed with mild detergent and deionized water, and sterilized with 70% ethanol.
Electrodes are designed to electroporate volumes ranging from 4mL to 60mL. Electrode assemblies contain parallel stainless steel electrodes, which produce homogeneous fields.