SP Bel-Art Stainless Steel Lab Spoon and Spatula, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F36711-0012
57952-250CS 933.23 USD
57952-250
SP Bel-Art Stainless Steel Lab Spoon and Spatula, Bel-Art Products, a part of SP
Spatulas
This large stainless steel lab spoon is 30.5 cm (12") long for reaching into larger bottles.
  • Stainless steel construction
  • Long handle provides extra reach
  • Two ends are spatula and spoon
One end is a flat spatula measuring 3.1 cm (1¼") wide; the spoon end is 3.7 cm (1½") wide and holds 5 ml. The connecting bar is 14 mm (½") wide.