Chemware® Micro Spatula, Teflon® Resin-Coated, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
D1069292
57949-033EA 61.06 USD
57949-033
Chemware® Micro Spatula, Teflon® Resin-Coated, Saint-Gobain Performance Plastics
Spatulas
Both ends have a durable Teflon® resin coating, which gives an inert surface for handling chemicals and prevents loss due to sticking.
  • Hayman style