Chemware® Laboratory Matting, PTFE, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
D1069328
57462-006EA 1269.64 USD
57462-006
Chemware® Laboratory Matting, PTFE, Saint-Gobain Performance Plastics
Surface Protectors Equipment Protectors
Ideal liner for sinks, ovens and freezers
Nonstick Teflon® resin mesh has openings of approximately 6.4mm (1/4"). Thickness is approximately 4.2mm (5/32"), permitting easy cutting to desired shape or size. Web structure is nonporous, preventing absorption of chemicals, stains, and odors. May be used at extreme temperatures of –240 to 316°C (–400° to 600°F). Use with Teflon® resin cord to create a chemically inert dipping basket.